Få mere at vide
Zopiclone, 1mg/ml in Acetonitrile, Lipomed
Artikelnummer. 30462877
Skift visning
Mængde:
1 ml
Artikelnummer. Mængde
30462877 1 ml
1 options
Artikelnummer. 30462877 Leverandør Lipomed Leverandørnr. LPMZOP921FB1LA
Lipomed delivers certified reference materials you can trust as neats, native solutions and stable isotope labelled solutions, with precise documentation and expert support. From forensic to clinical labs, choose Lipomed for confidence, compliance, and analytical excellence.
Zopiclone, 1mg/ml in Acetonitrile
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Zopiclone
CAS 43200-80-2
Analyt- eller komponentnavne Zopiclone
Forsendelsestilstand Room Temperature
Molekylær formel C17 H17 Cl N6 O3
Koncentration 1 mg/mL
Synonym Zopiclone
InChI formel InChI=1 S/C16H21NO2.ClH/c1-12(2)17-10-14(18)11-19-16-9-5-7-13-6-3-4-8-15(13)16;/h3-9,12,14,17-18 H,10-11H2,1-2H3;1 H
SMIL CN1CCN(CC1)C(=O)OC2N(C(=O)c3nccnc23)c4ccc(Cl)cn4
IUPAC navn [6-(5-chloropyridin-2-yl)-5-oxo-7 H-pyrrolo[3,4-b]pyrazin-7-yl] 4-methylpiperazine-1-carboxylate
Opløsningsmiddel eller Matrix Acetonitrile
Industry Type Pharmaceutical
Molekylvægt (g/mol) 388.81
Mængde 1 ml
Formel vægt 388.1051 g/mol