Få mere at vide
Zolpyridine (4-Methyl-N-(5-methyl-2-pyridinyl)benzamide), Mikromol™
Artikelnummer. 30050862
Skift visning
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30050862 25 mg
30050733 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30050862 Leverandør Mikromol Leverandørnr. MM0333.080025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Zolpyridine (4-Methyl-N-(5-methyl-2-pyridinyl)benzamide)
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Zolpyridine (4-Methyl-N-(5-methyl-2-pyridinyl)benzamide)
CAS 349122-64-1
Analyt- eller komponentnavne Zolpyridine (4-Methyl-N-(5-methyl-2-pyridinyl)benzamide)
Forsendelsestilstand Room Temperature
Molekylær formel C14 H14 N2 O
Certificeringer/compliance ISO 17025
Synonym Zolpyridine, 4-Methyl-N-(5-methyl-2-pyridinyl)benzamide, Zolpidem Related Compound C
InChI formel InChI=1S/C14H14N2O/c1-10-3-6-12(7-4-10)14(17)16-13-8-5-11(2)9-15-13/h3-9H,1-2H3,(H,15,16,17)
SMIL Cc1ccc(cc1)C(=O)Nc2ccc(C)cn2
IUPAC navn 4-methyl-N-(5-methylpyridin-2-yl)benzamide
Molekylvægt (g/mol) 226.27
Mængde 25 mg
Formel vægt 226.1106 g/mol