Få mere at vide
Zolpidem Phenyl-4-carboxylic Acid Ethyl Ester-d6, TRC
Artikelnummer. 30453939
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30453939 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30453939 Leverandør Toronto Research Chemicals Leverandørnr. TRCZ65002210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Zolpidem Phenyl-4-carboxylic Acid Ethyl Ester-d6
CAS 1216455-48-9
Molekylær formel C21H17D6N3O3
Mængde 10 mg
Synonym 4-[3-[2-(Dimethylamino)-2-oxoethyl]-6-methylimidazo[1,2-a]pyridin-2-yl]benzoic Acid Ethyl Ester-d6.
InChI formel InChI=1S/C21H23N3O3/c1-5-27-21(26)16-9-7-15(8-10-16)20-17(12-19(25)23(3)4)24-13-14(2)6-11-18(24)22-20/h6-11,13H,5,12H2,1-4H3/i3D3,4D3
SMIL [2H]C([2H])([2H])N(C(=O)Cc1c(nc2ccc(C)cn12)c3ccc(cc3)C(=O)OCC)C([2H])([2H])[2H]
IUPAC navn ethyl 4-[3-[2-[bis(trideuteriomethyl)amino]-2-oxoethyl]-6-methylimidazo[1,2-a]pyridin-2-yl]benzoate
Molekylvægt (g/mol) 371.47
Formel vægt 371.2116