Få mere at vide
Zolpidem-d6 6-Carboxylic Acid, TRC
Artikelnummer. 30450598
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30450598 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30450598 Leverandør Toronto Research Chemicals Leverandørnr. TRCZ65001210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Zolpidem-d6 6-Carboxylic Acid
CAS 1190014-62-0
Molekylær formel C19H13D6N3O3
Mængde 10 mg
InChI formel InChI=1S/C19H19N3O3/c1-12-4-6-13(7-5-12)18-15(10-17(23)21(2)3)22-11-14(19(24)25)8-9-16(22)20-18/h4-9,11H,10H2,1-3H3,(H,24,25)/i2D3,3D3
SMIL [2H]C([2H])([2H])N(C(=O)Cc1c(nc2ccc(cn12)C(=O)O)c3ccc(C)cc3)C([2H])([2H])[2H]
Molekylvægt (g/mol) 343.41
Formel vægt 343.1803