Få mere at vide
Zolpidem-d6 6-Carboxylic Acid Methyl Ester, TRC
Artikelnummer. 30451099
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30451099 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30451099 Leverandør Toronto Research Chemicals Leverandørnr. TRCZ6500271MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Zolpidem-d6 6-Carboxylic Acid Methyl Ester
CAS 1215767-20-6
Molekylær formel C20H15D6N3O3
Mængde 1 mg
Synonym 3-[2-(Dimethylamino)-2-oxoethyl]-2-(4-methylphenyl)-imidazo[1,2-a]pyridine-6-carboxylic Acid-d6 Methyl Ester
InChI formel InChI=1S/C20H21N3O3/c1-13-5-7-14(8-6-13)19-16(11-18(24)22(2)3)23-12-15(20(25)26-4)9-10-17(23)21-19/h5-10,12H,11H2,1-4H3/i2D3,3D3
SMIL [2H]C([2H])([2H])N(C(=O)Cc1c(nc2ccc(cn12)C(=O)OC)c3ccc(C)cc3)C([2H])([2H])[2H]
IUPAC navn methyl 3-[2-[bis(trideuteriomethyl)amino]-2-oxoethyl]-2-(4-methylphenyl)imidazo[1,2-a]pyridine-6-carboxylate
Molekylvægt (g/mol) 357.44
Formel vægt 357.196