Få mere at vide
Zolmitriptan-d6 N-Oxide, TRC
Artikelnummer. 30454642
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30454642 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30454642 Leverandør Toronto Research Chemicals Leverandørnr. TRCZ63901210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Zolmitriptan-d6 N-Oxide
CAS 1217618-32-0
Molekylær formel C16H15D6N3O3
Mængde 10 mg
Synonym (4S)-4-[[3-[2-(Dimethyl-d6-oxidoamino)ethyl]-1H-indol-5-yl]methyl-2-oxazolidinone
InChI formel InChI=1S/C16H21N3O3/c1-19(2,21)6-5-12-9-17-15-4-3-11(8-14(12)15)7-13-10-22-16(20)18-13/h3-4,8-9,13,17H,5-7,10H2,1-2H3,(H,18,20)/t13-/m0/s1/i1D3,2D3
SMIL O=C1OC[C@@H](N1)CC2=CC(C(CCN(C([2H])([2H])[2H])(C([2H])([2H])[2H])=O)=CN3)=C3C=C2
Molekylvægt (g/mol) 309.39
Formel vægt 309.196