Få mere at vide
Zoledronic-d3 Acid, TRC
Artikelnummer. 30451606
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30451606 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30451606 Leverandør Toronto Research Chemicals Leverandørnr. TRCZ6235041MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Zoledronic-d3 Acid
CAS 1134798-26-7
Molekylær formel C5H7D3N2O7P2
Mængde 1 mg
Synonym P,P'-[1-Hydroxy-2-(1H-imidazol-1-yl-d3-2,4,5-d3)ethylidene]bis-phosphonic Acid
InChI formel InChI=1S/C5H10N2O7P2/c8-5(15(9,10)11,16(12,13)14)3-7-2-1-6-4-7/h1-2,4,8H,3H2,(H2,9,10,11)(H2,12,13,14)/i1D,2D,4D
SMIL O=P(O)(O)C(CN1C([2H])=NC([2H])=C1[2H])(O)P(O)(O)=O
Molekylvægt (g/mol) 275.11
Formel vægt 275.0152