Få mere at vide
Zofenopril Calcium Salt, TRC
Artikelnummer. 30138585
Skift visning
Mængde:
5 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30138585 10 mg
30005759 5 mg
30005807 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30138585 Leverandør Toronto Research Chemicals Leverandørnr. TRCZ58700010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 81938-43-4
Molekylær formel 2 C22 H22 N O4 S2 . Ca
Molekylvægt (g/mol) 897.17
Synonym L-Proline, 1-[(2S)-3-(benzoylthio)-2-methyl-1-oxopropyl]-4-(phenylthio)-, calcium salt (2:1), (4S)-, L-Proline, 1-[3-(benzoylthio)-2-methyl-1-oxopropyl]-4-(phenylthio)-, calcium salt, [1(R*),2α,4α]-, SQ 26991, Zofenil, Zofenopril calcium, Zoprace
IUPAC navn calcium;(2S,4S)-1-[(2S)-3-benzoylsulfanyl-2-methylpropanoyl]-4-phenylsulfanylpyrrolidine-2-carboxylate
SMIL [Ca+2].C[C@H](CSC(=O)c1ccccc1)C(=O)N2C[C@H](C[C@H]2C(=O)[O-])Sc3ccccc3.C[C@H](CSC(=O)c4ccccc4)C(=O)N5C[C@H](C[C@H]5C(=O)[O-])Sc6ccccc6
Tekniske data
Kemisk navn eller materiale Zofenopril Calcium Salt
InChI formel InChI=1S/2C22H23NO4S2.Ca/c2*1-15(14-28-22(27)16-8-4-2-5-9-16)20(24)23-13-18(12-19(23)21(25)26)29-17-10-6-3-7-11-17;/h2*2-11,15,18-19H,12-14H2,1H3,(H,25,26);/q;;+2/p-2/t2*15-,18+,19+;/m11./s1
Mængde 10 mg
Formel vægt 896.1606