Beskrivelse
Antibody detects endogenous levels of total ZNF785.
The function of ZNF785 remains unknown.
Tekniske data
Tekniske data
| Antigen | ZNF785 |
| Applikationer | Western Blot, Immunocytochemistry |
| Klassifikation | Polyclonal |
| Koncentration | 1 mg/mL |
| Konjugeret | Unconjugated |
| Formulering | PBS with 50% glycerol and 0.02% sodium azide |
| Gene | ZNF785 |
| Genadgangsnr. | A8K8V0 |
| Gene Alias | Zinc finger protein 785; zinc finger protein 785; LOW QUALITY PROTEIN: zinc finger protein 785; ZNF785 |
| Gensymboler | ZNF785 |
| Vis mere |