missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF358 Antibody (PCRP-ZNF358-1A6), Alexa Fluor™ 405, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-14233AF405
Additional Details : Weight : 0.00970kg
Description
ZNF358 Monoclonal antibody specifically detects ZNF358 in Human samples. It is validated for Western Blot, Flow Cytometry, ImmunofluorescenceSpecifications
ZNF358 | |
Monoclonal | |
Alexa Fluor 405 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
140467 | |
Store at 4°C in the dark. | |
Purified |
Western Blot, Flow Cytometry, Immunofluorescence | |
PCRP-ZNF358-1A6 | |
Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence | |
hKID3, KID3Kidney, ischemia, and developmentally-regulated protein 3, KRAB-zinc finger protein synten, zinc finger protein 354C | |
Recombinant full-length human ZNF358protein (Uniprot: Q9NW07) | |
0.1 mL | |
Primary | |
Human | |
Antibody | |
IgG2c |