missing translation for 'onlineSavingsMsg'
Få mere at vide

ZM39923 hydrochloride, MedChemExpress

Product Code. 16431467
Encompass_Preferred
Change view
Click to view available options
Mængde:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
16431467 50 mg
16425258 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16431467 Supplier Medchem Express Supplier No. HY12589
 Begrænset vare
Forventet forsendelse: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress ZM39923 hydrochloride is a JAK3 inhibitor, with a pIC50 of 7.1; ZM39923 hydrochloride also potently inhibits tissue transglutaminase (TGM2) with an IC50 of 10 nM.

ZM39923 hydrochloride is a JAK3 inhibitor, with a pIC50 of 7.1

Specifications

Kemisk navn eller materiale ZM39923 hydrochloride
Fysisk form Solid
CAS 1021868-92-7
Farve White
Mængde 50 mg
Renhedsgrad noter Research
Molekylær formel C23H26ClNO
Til brug med (applikation) Cancer-Kinase/protease
Opløselighedsinformation DMSO : ≥ 47 mg/mL (127.75 mM) ∣H2O : 1 mg/mL (2.72 mM; Need ultrasonic)
SMIL O=C(C1=CC=C2C=CC=CC2=C1)CCN(C(C)C)CC3=CC=CC=C3.[H]Cl
Molekylvægt (g/mol) 367.91
Formel vægt 367.91
Procent renhed 99.86%
Grad Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.