Få mere at vide
Tocris Bioscience™ ZLN 024 hydrochloride
Mængde:
10 mg
50 mg
Beskrivelse
ZLN 024 hydrochloride is an allosteric AMP-activated protein kinase (AMPK) activator; allosterically stimulates active AMPK heterotrimers, increasing the activity of α1β1γ1 by 1.5-fold and α2β1γ1 by 1.7-fold (EC50 values are 0.42 and 0.95 μM, respectively). Inhibits dephosphorylation of AMPK Thr-172 by PP2Cα. Stimulates glucose uptake and fatty acid oxidation without increasing the ADP/ATP ratio. Also decreases fatty acid synthesis and glucose output in primary hepatocytes. Improves glucose tolerance and decreases cholesterol content in db/db mice.
Tekniske data
Tekniske data
| Kemisk navn eller materiale | 2-[[2-(2-Bromo-4-methylphenoxy)ethyl]thio]pyrimidine hydrochloride |
| Mængde | 50 mg |
| Mål | AMPK Activators |
| Molekylær formel | C13H13BrN2OS.HCl |
| Formel vægt | 361.69 |
| Renhed | >99% |