Få mere at vide
ZK 159222, TRC
Artikelnummer. 30451020
Skift visning
Mængde:
0.5 mg
5 mg
Pakningsstørrelse:
0.5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30451020 0.5 mg 0.5mg
30453588 5 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30451020 Leverandør Toronto Research Chemicals Leverandørnr. TRCZ4858000.5MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 156965-15-0
Molekylær formel C32H48O5
Molekylvægt (g/mol) 512.72
Synonym ZK 159222, 1-[(1α,3β,5Z,7E,22E,24R)-1,3,24-Trihydroxy-9,10-secochola-5,7,10(19),22-tetraen-24-yl]-cyclopropanecarboxylic Acid Butyl Ester
IUPAC navn butyl 1-[(E,1R,4R)-4-[(1R,3aS,4E,7aR)-4-[(2Z)-2-[(3S,5R)-3,5-dihydroxy-2-methylidenecyclohexylidene]ethylidene]-7a-methyl-2,3,3a,5,6,7-hexahydro-1H-inden-1-yl]-1-hydroxypent-2-enyl]cyclopropane-1-carboxylate
SMIL CCCCOC(=O)C1(CC1)[C@H](O)\C=C\[C@@H](C)[C@H]2CC[C@H]3\C(=C\C=C/4\C[C@@H](O)C[C@H](O)C4=C)\CCC[C@]23C
Tekniske data
Kemisk navn eller materiale 1-[(1alpha,3beta,5Z,7E,22E,24R)-1,3,24-Trihydroxy-9,10-secochola-5,7,10(19),22-tetraen-24-yl]-cyclopropanecarboxylic Acid Butyl Ester
Mængde 0.5 mg
InChI formel InChI=1S/C32H48O5/c1-5-6-18-37-30(36)32(16-17-32)29(35)14-9-21(2)26-12-13-27-23(8-7-15-31(26,27)4)10-11-24-19-25(33)20-28(34)22(24)3/h9-11,14,21,25-29,33-35H,3,5-8,12-13,15-20H2,1-2,4H3/b14-9+,23-10+,24-11-/t21-,25-,26-,27+,28+,29-,31-/m1/s1
Formel vægt 512.3502