Learn More
RETSCH Zirconium Oxide Grinding Jars
Product Code. 15390964 Shop All Retsch Products
Change view
Click to view available options
For Use With (Equipment):
For CryoMill
For EMAX
For MM 200 Mixer Mill
Capacity (Metric):
10 mL
25 mL
50 mL
125 mL
Unit Size:
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Capacity (Metric) Unit Size
15390964 For EMAX 50 mL Each
15380964 For EMAX 125 mL Each
14841631 For CryoMill 25 mL Each
10623011 For MM 200 Mixer Mill 25 mL Each
10636141 For MM 200 Mixer Mill 10 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 15390964 Supplier RETSCH Supplier No. 01.462.0312
  • ONLINE EXCLUSIVE PRICE
    51585.00 DKK
    46233.03 DKK / Pakke med 1
    Spare 5351.97 DKK
    10% Slukket
Forventet forsendelse: 08-02-2027
Spare Grinding Jar
  • Accessory
Specifications
Product Type Grinding Jar
Capacity (Metric) 50 mL
For Use With (Equipment) For EMAX
Material Zirconium Oxide