Få mere at vide
Ziprasidone-d8, TRC
Artikelnummer. 30454470
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30454470 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30454470 Leverandør Toronto Research Chemicals Leverandørnr. TRCZ4850021MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Ziprasidone-d8
CAS 1126745-58-1
Renhedsgrad noter HPLC
Molekylær formel C21 D8 H13 Cl N4 O S
Mængde 1 mg
Synonym 5-[2-[4-(1,2-Benzisothiazol-3yl)-1-(piperazinyl-d8)]ethyl]-6-chloro-1,3-dihydro-2H-indol-2-one
InChI formel InChI=1S/C21H21ClN4OS/c22-17-13-18-15(12-20(27)23-18)11-14(17)5-6-25-7-9-26(10-8-25)21-16-3-1-2-4-19(16)28-24-21/h1-4,11,13H,5-10,12H2,(H,23,27)/i7D2,8D2,9D2,10D2
SMIL [2H]C1([2H])N(CCc2cc3CC(=O)Nc3cc2Cl)C([2H])([2H])C([2H])([2H])N(c4nsc5ccccc45)C1([2H])[2H]
IUPAC navn 5-[2-[4-(1,2-benzothiazol-3-yl)-2,2,3,3,5,5,6,6-octadeuterio-piperazin-1-yl]ethyl]-6-chloro-indolin-2-one
Molekylvægt (g/mol) 420.98
Formel vægt 420.1627
Procent renhed >95