Learn More
Chemsplash Zip Labcoat T5/6 Fabric, Elastic Cuff
GREENER_CHOICE
Lab Coat is a 2/3 length coat with a collar and zip closure and elasticated wrists. It is anti-static to meet with EN1149-5:2008, lint free and suitable for clean room applications.
2405.00 DKK
Specifications
Product Type Lab Coat
Gender Unisex
Material Microporous Composite Fabric
Color White
Closure Type Zipper
View More Specs
Products 6
Product Code Brand Size Price Quantity & Availability  
Product Code Brand Size Price Quantity & Availability  
30058202
GREENER_CHOICE
View Documents
Chemsplash
2549.400.004
Small
2405.00 DKK
Pack of 50
Only null left
Estimated Shipment: 25-07-2025
Add to basket
30058450
GREENER_CHOICE
View Documents
Chemsplash
2549.400.005
Medium
2405.00 DKK
Pack of 50
Only null left
Estimated Shipment: 25-07-2025
Add to basket
30058447
GREENER_CHOICE
View Documents
Chemsplash
2549.400.006
Large
2405.00 DKK
Pack of 50
Only null left
Estimated Shipment: 25-07-2025
Add to basket
30058301
GREENER_CHOICE
View Documents
Chemsplash
2549.400.007
X-Large
2405.00 DKK
Pack of 50
Only null left
Estimated Shipment: 25-07-2025
Add to basket
30058165
GREENER_CHOICE
View Documents
Chemsplash
2549.400.008
2X-Large
2405.00 DKK
Pack of 50
Only null left
Estimated Shipment: 25-07-2025
Add to basket
30058354
GREENER_CHOICE
View Documents
Chemsplash
2549.400.009
3X-Large
2405.00 DKK
Pack of 50
Only null left
Estimated Shipment: 25-07-2025
Add to basket
Description
Description
Chemsplash Lab Coat
  • Chemsplash 55GSM Type PB 6B Fabric
  • Zip Fasten Front with Adhesive Flap Cover
  • Elasticated Cuffs
  • Fold Over Collar
  • Silicone and Latex Free
  • Anti-Static Material
  • Non-Linting
  • PFAS Free
  • Conforms to ISO 14644-1:2015 Cleanroom 5 - 9
Description: The Chemsplash Lab Coat is made from Chemsplash fabric, which is soft and microporous. It is strong and resistant to chemical splash and dust. This lab coat is a 2/3 length coat with a collar, zip closure, and elasticated wrists. It is anti-static to meet EN1149-5:2008, lint-free, and suitable for cleanroom applications. Conforms to ISO 14644-1:2015 Cleanroom 5 - 9. Chemsplash accessories can be used to protect the wearer from the environment or the environment from the wearer. The range has been designed to allow for maximum comfort and wearability.
Specifications
Specifications
Lab Coat
Microporous Composite Fabric
Zipper
Single-use
Liquids
Yes
Class 100-100,000 (ISO 5-8)
Unisex
White
Elastic
Non-sterile
None
Autoclavable
Documents
Product Certifications
Product Content Correction
Product Title
Chemsplash Zip Labcoat T5/6 Fabric, Elastic Cuff