Learn More
Zincon Sodium Salt, MP Biomedicals™
Catalog Number p-4679581
Click to view available options
Quantity:
5 g
62625-22-3
C20H16N4NaO6S
463.42
IABRINWJUBAIIE-JJECXDOKSA-N
2-Carboxy-2'-hydroxy-5'-sulfoformazylbenzene
131856391
2-[2-[(Z)-N-[(Z)-(6-oxo-3-sulfocyclohexa-2,4-dien-1-ylidene)amino]-C-phenylcarbonimidoyl]hydrazinyl]benzoic acid;sodium
C1=CC=C(C=C1)C(=NN=C2C=C(C=CC2=O)S(=O)(=O)O)NNC3=CC=CC=C3C(=O)O.[Na]
62625-22-3
C20H16N4NaO6S
463.42
IABRINWJUBAIIE-JJECXDOKSA-N
2-Carboxy-2'-hydroxy-5'-sulfoformazylbenzene
131856391
2-[2-[(Z)-N-[(Z)-(6-oxo-3-sulfocyclohexa-2,4-dien-1-ylidene)amino]-C-phenylcarbonimidoyl]hydrazinyl]benzoic acid;sodium
C1=CC=C(C=C1)C(=NN=C2C=C(C=CC2=O)S(=O)(=O)O)NNC3=CC=CC=C3C(=O)O.[Na]