missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zinc fluoride, 98%, ACROS Organics™
345.00 DKK - 1885.00 DKK
Chemical Identifiers
CAS | 7783-49-5 |
---|---|
Molecular Formula | F2Zn |
Molecular Weight (g/mol) | 103.38 |
MDL Number | MFCD00011298 |
InChI Key | BHHYHSUAOQUXJK-UHFFFAOYSA-L |
Synonym | zinc ii fluoride, zinc 2+ ion difluoride, acmc-1bhy1, ksc379c0l, zinc fluoride 50g, zinc, ion zn2+ difluoride |
PubChem CID | 10129912 |
IUPAC Name | zinc;difluoride |
SMILES | [F-].[F-].[Zn+2] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10009061
|
Acros Organics
363910250 |
25g | Plastic bottle |
345.00 DKK
25g |
In stock, ready to ship: 1 | Please sign in to purchase this item. Need a web account? Register with us today! | |||
10510362
|
Acros Organics
363912500 |
250g | Plastic bottle |
1885.00 DKK
250g |
Estimated Shipment 15-03-2021 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Chemical Identifiers
7783-49-5 | |
103.38 | |
BHHYHSUAOQUXJK-UHFFFAOYSA-L | |
10129912 | |
[F-].[F-].[Zn+2] |
F2Zn | |
MFCD00011298 | |
zinc ii fluoride, zinc 2+ ion difluoride, acmc-1bhy1, ksc379c0l, zinc fluoride 50g, zinc, ion zn2+ difluoride | |
zinc;difluoride |
Specifications
Zinc fluoride | |
7783-49-5 | |
F2Zn | |
MFCD00011298 | |
15, 10335 | |
Solubility in water: slightly soluble. Other solubilities: freely soluble in hcl,hno3 and nh4oh,insoluble in alcohol | |
[F-].[F-].[Zn+2] | |
103.38 | |
103.38 | |
98% | |
Solubility in water: slightly soluble. Other solubilities: freely soluble in hcl, hno3 and nh4oh, insoluble in alcohol | |
1500.0°C | |
872.0°C to 910.0°C |
98% | |
98% | |
ZnF2 | |
06,514 | |
zinc ii fluoride, zinc 2+ ion difluoride, acmc-1bhy1, ksc379c0l, zinc fluoride 50g, zinc, ion zn2+ difluoride | |
BHHYHSUAOQUXJK-UHFFFAOYSA-L | |
zinc;difluoride | |
10129912 | |
Powder or Crystals | |
Plastic bottle | |
61.7 to 64.8 % | |
Brown to White | |
25g |