Få mere at vide
Zinc Dimethyldithiocarbamate, TRC
Artikelnummer. 30451471
Skift visning
Mængde:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30451471 10 g
30456088 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30451471 Leverandør Toronto Research Chemicals Leverandørnr. TRCZ43965010G
Denne vare kræver en prisopdatering, inden den kan bestilles.
Anmod om en pris på denne vare her.
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 137-30-4
Molekylær formel C6 H12 N2 S4 Zn
Molekylvægt (g/mol) 305.84
Synonym Zinc, bis(N,N-dimethylcarbamodithioato-κS,κS')-, (T-4)-, Zinc, bis(dimethylcarbamodithioato-S,S')-, (T-4)-, Zinc, bis(dimethylcarbamodithioato-κS,κS')-, (T-4)- (9CI), Zinc, bis(dimethyldithiocarbamato)- (8CI), (T-4)-Bis(N,N-dimethylcarbamodithioato-κS,κS')zinc, Carbamodithioic acid, dimethyl-, zinc complex, Aaprotect, Aavolex, Aazira, Accel PZ, Accelerator L, Accelerator MZ Powder, Accelerator MZX, Accelerator PZ, Aceto ZDED, Aceto ZDMD, Alcobam ZM, Ancazate ME, Bis(dimethyldithiocarbamato)zinc, Carbamodithioic acid, dimethyl-, zinc salt, Carbazinc, Corona Corozate, Corozate, Crittam, Cuman, Cuman L, Cymate, Eptac 1, Fuclasin, Fuclasin-Ultra, Fuklasin, Fulasin, Hermat ZDM, Hexazir, Karbam White, Methasan, Methazate, Methyl Ziram, Methyl zimate, Methyl zineb, Mezene, Milbam, Molurame, Mycronil, NSC 737, Nocceler PZ, Octocure ZDM 50, Orchard brand Ziram, PZ, Perkacit ZDMC, Pomarzol Z-forte, Rhenogran ZDMC 80, Rhodiacid, Rodisan, Sanceler PZ, Soxinal PZ, Soxinol PZ, Trikagol, Ultra Zinc DMC, Vancide MZ 96, Vulcacure ZM, Vulkacit L, Vulkacite L, Z 75, Z-C Spray, ZDMC, ZDMC 80, Zarlate, Zerlate, Zimate, Zinc bis(dimethyldithiocarbamate), Zinc dimethyldithiocarbamate, Ziradin, Ziram, Zirberk, Ziride, Zirol, Zirthane, ZnMDC
IUPAC navn bis(dimethylcarbamothioylsulfanyl)zinc
SMIL CN(C)C(=S)S[Zn]SC(=S)N(C)C
Tekniske data
Kemisk navn eller materiale Ziram
Mængde 10 g
InChI formel InChI=1S/2C3H7NS2.Zn/c2*1-4(2)3(5)6;/h2*1-2H3,(H,5,6);/q;;+2/p-2
Formel vægt 303.9175