Få mere at vide
Zinc Dibutyldithiocarbamate, TRC
Artikelnummer. 30012105
Skift visning
Mængde:
500 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30012105 1 g
30008686 5 g
30005795 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30012105 Leverandør Toronto Research Chemicals Leverandørnr. TRCZ2178601G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 136-23-2
Molekylær formel C18H36N2S4Zn
Molekylvægt (g/mol) 474.12
Synonym (T-4)-Bis(dibutylcarbamodithioato-κS,κS′)zinc (ACI), Carbamodithioic acid, dibutyl-, zinc complex (ZCI), Zinc, bis(dibutylcarbamodithioato-S,S′)-, (T-4)- (ZCI), Zinc, bis(dibutyldithiocarbamato)- (7CI, 8CI), (Dibutyldithiocarbamato)zinc(II), Accel BZ, Accelerator BZ, Aceto ZDBD, Antioxidant BZ, Bis(dibutyldithiocarbamato)zinc, Bis(N,N-dibutyldithiocarbamato)zinc, Bostex 497B, Butazate, Butazate 50D, Butyl zimate, Butyl ziram, BZ, BZ 75, BZ-P, Carbamodithioic acid, dibutyl-, zinc salt, Carbamodithioic acid, N,N-dibutyl-, zinc salt (2:1), Dibutyldithiocarbamic acid zinc salt, Nocceler BZ, Nocceler BZ-G, Nocceler BZ-P, Nocceler BZ-S, NSC 36548, NSC 3880, Perkacit ZDBC, Rhenocure LDB/C, Rhenogran ZBDC 80, Rhenogran ZDBC 75, Rhenogran ZDBC 80, Rhexon ZDBC 75, Sanceler BZ, Soxinol BZ, T 202, T 202 (antioxidant), T 353, Transcure ZDB 50, Vulcacure ZB, Vulcafor ZDBC, Vulkacit LDB, Vulkacit LDS, ZDBC, ZDBC 50, ZDBC 75, ZDBC 80, Zinc bis(dibutyldithiocarbamate), Zinc bis(N,N-dibutyldithiocarbamate), Zinc di-n-butylthiocarbamate, Zinc bis(N,N-dibutyldithiocarbamate), Zinc bis(dibutyldithiocarbamate), Zinc dibutyldithiocarbamate, Zinc N,N-dibutyldithiocarbamate, ZnBDC
IUPAC navn bis((dibutylcarbamothioyl)thio)zinc
SMIL CCCCN(CCCC)C(S[Zn]SC(N(CCCC)CCCC)=S)=S
Tekniske data
Kemisk navn eller materiale Zinc Dibutyldithiocarbamate
Mængde 1 g
InChI formel InChI=1S/2C9H19NS2.Zn/c2*1-3-5-7-10(9(11)12)8-6-4-2;/h2*3-8H2,1-2H3,(H,11,12);/q;;+2/p-2
Formel vægt 472.11