missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zinc citrate dihydrate, Reagent Grade, Zn 31% min, Thermo Scientific Chemicals
234.50 DKK valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
250 g
1 kg
Description
Zinc citrate dihydrate is used in dental care products such as toothpastes, mouthwashes and chewing gums due to its antimicrobial and anti-inflammatory effects. It used in used in food supplements, functional foods and beverages. It is used to formulated in pharmaceuticals, and foods as a zinc supplement. It used as intermediate in organic synthesis, agrochemicals and dyestuff.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 334°C (decomposition) |
Quantity | 250 g |
Solubility Information | Soluble in dilute mineral acids and alkali hydroxides. Slightly soluble in water |
Formula Weight | 610.35 (574.32 Anhydrous) |
Grade | Reagent |
Physical Form | Powder |
Chemical Name or Material | Zinc citrate dihydrate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'