Learn More
BRAND™ Zinc Alloy Burette Clamps
PVC-coated clamps provide excellent non-slip, scratch-resistant grip.
Brand: BRAND™ 16515
Additional Details : Weight : 0.18000kg
Description
- Zinc alloy
- Nickel-plated
- PVC-coated grips
Specifications
For 1 Burette | |
Zinc Alloy-plated | |
1 Each | |
Buret Clamp |