missing translation for 'onlineSavingsMsg'
Learn More

Zinc Acetate Dihydrate, Reagent Grade, Honeywell™

Catalog Number 15608210
Click to view available options
Quantity:
500 g
1 kg
Packaging:
Poly Bottle
This item is not returnable. View return policy
5970-45-6
C4H10O6Zn
219.498
MFCD00066961
BEAZKUGSCHFXIQ-UHFFFAOYSA-L
zinc acetate dihydrate, zinc diacetate dihydrate, zinc ii acetate dihydrate, unii-fm5526k07a, acetic acid zinc salt, zinc acetate usp, acetic acid, zinc salt, dihydrate, zinc acetate, dihydrate, galzin tn
2724192
zinc;diacetate;dihydrate
CC(=O)[O-].CC(=O)[O-].O.O.[Zn+2]
This item is not returnable. View return policy
5970-45-6
C4H10O6Zn
219.498
MFCD00066961
BEAZKUGSCHFXIQ-UHFFFAOYSA-L
zinc acetate dihydrate, zinc diacetate dihydrate, zinc ii acetate dihydrate, unii-fm5526k07a, acetic acid zinc salt, zinc acetate usp, acetic acid, zinc salt, dihydrate, zinc acetate, dihydrate, galzin tn
2724192
zinc;diacetate;dihydrate
CC(=O)[O-].CC(=O)[O-].O.O.[Zn+2]

Reagent Grade

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY
Melting Point ∼100°C
Color Gray
pH 6.0 to 8.0 (at 20°C)
Quantity 1 kg
Linear Formula Zn(CH3COO)2 · 2H2O
UN Number UN3077
Beilstein 3732513
Formula Weight 219.51g/mol
Odor Vinegar-like
Packaging Poly Bottle
Appearance Crystalline
Density 1.750g/cu.cm at 20°C
Chemical Name or Material Zinc acetate dihydrate
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Zinc acetate dihydrate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H318-Causes serious eye damage.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
missing translation for 'delimitedSupplementalStatement'
  • MIXTURE LIST-Contains : Zinc acetate-2-hydrate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'