Få mere at vide
Zinc 2-Mercaptobenzothiazole, TRC
Artikelnummer. 30138337
Skift visning
Mængde:
5 g
10 g
50 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30138337 50 g
30133297 5 g
30003772 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30138337 Leverandør Toronto Research Chemicals Leverandørnr. TRCZ70156850G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 155-04-4
Molekylær formel C14H8N2S4Zn
Molekylvægt (g/mol) 397.85
IUPAC navn zinc;1,3-benzothiazole-2-thiolate
SMIL [Zn+2].[S-]c1nc2ccccc2s1.[S-]c3nc4ccccc4s3
Tekniske data
Kemisk navn eller materiale Zinc 2-Mercaptobenzothiazole
Mængde 50 g
InChI formel InChI=1S/2C7H5NS2.Zn/c2*9-7-8-5-3-1-2-4-6(5)10-7;/h2*1-4H,(H,8,9);/q;;+2/p-2
Formel vægt 395.8862