Få mere at vide
Zimeldine-d6, TRC
Artikelnummer. 30452114
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30452114 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30452114 Leverandør Toronto Research Chemicals Leverandørnr. TRCZ43500210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Zimeldine-d6
CAS 1185239-75-1
Molekylær formel C16H11D6BrN2
Mængde 10 mg
Synonym (Z)-3-(4-Bromophenyl)-(N,N-dimethyl-d6)-3-(3-pyridinyl)-2-propen-1-amine, Zimelidine-d6
InChI formel InChI=1S/C16H17BrN2/c1-19(2)11-9-16(14-4-3-10-18-12-14)13-5-7-15(17)8-6-13/h3-10,12H,11H2,1-2H3/b16-9-/i1D3,2D3
SMIL [2H]C([2H])([2H])N(C\C=C(\c1ccc(Br)cc1)/c2cccnc2)C([2H])([2H])[2H]
IUPAC navn (Z)-3-(4-bromophenyl)-3-pyridin-3-yl-N,N-bis(trideuteriomethyl)prop-2-en-1-amine
Molekylvægt (g/mol) 323.26
Formel vægt 322.0952