Få mere at vide
Zileuton-d4 (major), TRC
Artikelnummer. 30449621
Skift visning
Mængde:
25 mg
Artikelnummer. Mængde
30449621 25 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30449621 Leverandør Toronto Research Chemicals Leverandørnr. TRCZ42000225MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Zileuton-d4
CAS 1189878-76-9
Molekylær formel C11 D4 H8 N2 O2 S
Mængde 25 mg
Synonym 1-[1-(1-Benzothiophen-2-yl)-1,2,2,2-tetradeuterioethyl]-1-hydroxyurea, N-(1-Benzo[b]thien-2-ylethyl-1,2,2,2-d4)-N-hydroxyurea, 1-[1-(1-Benzothiophen-2-yl)ethyl-1,2,2,2-d4]-1-hydroxyurea, (±)-Zileuton-d4 (ethyl-d4), Zileuton-d4, Zileuton-D4 (ethyl-1,2,2,2-D4)
InChI formel InChI=1S/C11H12N2O2S/c1-7(13(15)11(12)14)10-6-8-4-2-3-5-9(8)16-10/h2-7,15H,1H3,(H2,12,14)/i1D3,7D
SMIL [2H]C([2H])([2H])C([2H])(N(O)C(=O)N)c1cc2ccccc2s1
IUPAC navn 1-[1-(1-benzothiophen-2-yl)-1,2,2,2-tetradeuterioethyl]-1-hydroxyurea
Molekylvægt (g/mol) 240.3148
Formel vægt 240.0871