Få mere at vide
(±)-Zileuton-d4 (ethyl-d4), CDN
Artikelnummer. 30320916
Skift visning
Mængde:
0.005 g
0.01 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30320916 0.01 g
30317757 0.005 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30320916 Leverandør CDN Leverandørnr. CDND78570.01G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Zileuton-d4
CAS 1189878-76-9
Molekylær formel C11 D4 H8 N2 O2 S
Mængde 0.01 g
Synonym 1-[1-(1-Benzothiophen-2-yl)-1,2,2,2-tetradeuterioethyl]-1-hydroxyurea, N-(1-Benzo[b]thien-2-ylethyl-1,2,2,2-d4)-N-hydroxyurea, 1-[1-(1-Benzothiophen-2-yl)ethyl-1,2,2,2-d4]-1-hydroxyurea, (±)-Zileuton-d4 (ethyl-d4), Zileuton-d4, Zileuton-D4 (ethyl-1,2,2,2-D4)
InChI formel InChI=1 S/C11H12N2O2S/c1-7(13(15)11(12)14)10-6-8-4-2-3-5-9(8)16-10/h2-7,15 H,1H3,(H2,12,14)/i1D3,7 D
SMIL [2 H]C([2 H])([2 H])C([2 H])(N(O)C(=O)N)c1cc2ccccc2s1
IUPAC navn 1-[1-(1-benzothiophen-2-yl)-1,2,2,2-tetradeuterioethyl]-1-hydroxyurea
Molekylvægt (g/mol) 240.3148
Formel vægt 240.0871 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity