missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZHX3 Antibody (PCRP-ZHX3-1D11), Alexa Fluor™ 405, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-14225AF405
Additional Details : Weight : 0.00970kg
Description
ZHX3 Monoclonal antibody specifically detects ZHX3 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, ImmunofluorescenceSpecifications
ZHX3 | |
Monoclonal | |
Alexa Fluor 405 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
23051 | |
Store at 4°C in the dark. | |
Purified |
Western Blot, Flow Cytometry, ELISA, Immunofluorescence | |
PCRP-ZHX3-1D11 | |
Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence | |
FLJ93221, Sp38, zona pellucida binding protein, zona pellucida-binding protein 1, ZPBP1zona pellucida binding protein 1 | |
Recombinant full-length human ZHX3 protein (Uniprot: Q9H4I2) | |
0.1 mL | |
Primary | |
Human | |
Antibody | |
IgG2b |