Learn More
3M™ Zeta Plus™ BC BC0025S90ZB08A Capsule Filter
3M™ Zeta Plus™ BC BC0025S90ZB08A Capsule Filter made of a highly porous carbonaceous material that is characterised by a large internal surface area providing exceptional adsorptive properties.
Brand:  3M™ 7100102763
Additional Details : Weight : 0.34620kg
Product Code. 15979471
  • 4450.00 DKK / Pack of 4
Estimated Shipment: 10-09-2024
to see stock.
Description
Description
  • Less fire risk and almost no carbon
  • After initial flush of the cartridge, there is minimal release of carbon fines in the filtered solution
  • Decreased process time because of constant flow rate filtration and efficiency
  • No product rework because of consistent performance
  • Activated carbon is fixed and the flow is forced through the matrix that increases efficiency
  • A single step is used as decolourisation is combined with the filtration step Reduced costs in solvent recovery
Specifications
Specifications
3 L/min
80°C
Filter Capsule