Learn More
ZEOprep™ Classic, (HYD) Moisture Content 4 to 6%
GREENER_CHOICE
ZEOprep™ is a packing material providing fast, effective, and reproducible separations. Irregular shaped silica gel is used in Gravity, Flash, and Medium Pressure Chromatography. ZEOprep HYD has a moisture content of 4 to 6% for processes better suited to a more hydrated silica.
616.00 DKK - 2645.00 DKK
Specifications
Pore Size 60 Ă…
Technique Chromatography
Grade Moisture content 4 to 6%
Type Silica Gel
Material Silica Gel
View More Specs
Products 4
Product Code Brand Description Price Quantity Availability  
Product Code Brand Description Price Quantity Availability  
30048982
GREENER_CHOICE
View Documents
ZEOprep
302103
40 to 63 ÎĽm, 5 kg 40 to 63 ÎĽm, 5 kg
2645.00 DKK
5kg
View alternative products
Save Up to 
»
30048953
GREENER_CHOICE
View Documents
ZEOprep
302104
60 to 200 ÎĽm, 5 kg 60 to 200 ÎĽm, 5 kg
2350.00 DKK
5kg
View alternative products
Save Up to 
»
30048990
GREENER_CHOICE
View Documents
ZEOprep
300211
60 to 200 ÎĽm, 1 kg 60 to 200 ÎĽm, 1 kg
616.00 DKK
1kg
View alternative products
Save Up to 
»
30049010
GREENER_CHOICE
View Documents
ZEOprep
300214
40 to 63 ÎĽm, 1 kg 40 to 63 ÎĽm, 1 kg
676.00 DKK
1kg
View alternative products
Save Up to 
»
Description
Description
  • Hydrated silica moisture content 4 to 6%
  • Robust silica for a wide variety of applications
  • Irregular Amorphous Silica Gel
  • High Purity
  • European Manufacture
  • Swiss accuracy and consistency
  • Excellent batch to batch reproducability
  • ISO Certification
  • High Chemical and Mechanical Stability
  • Narrow Particles Size Distribution
  • 20 to 500 ÎĽm particles size range
  • Available in 60 A or 90 A pore size
  • High loadability
  • Selection of pack sizes available
  • Suitable for gravity, flash, prep and process scale
Specifications
Specifications
60 Ă…
Moisture content 4 to 6%
Silica Gel
Irregular
Chromatography
Silica Gel
Bare Silica
Documents
Product Certifications
Product Content Correction
Product Title
ZEOprep™ Classic, (HYD) Moisture Content 4 to 6%