missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
STEMCELL Technologies Zebularine
Beskrivelse
- Zebularine, a cytidine analog, is a DNA methylation inhibitor that acts by forming a covalent complex with DNA methyltransferases (DNMTs; Zhou et al.)
- Induces cardiomyocyte differentiation from rat bone marrow mesenchymal stem cells (MSCs; Naeem et al.)
- Reduces proliferation of the cancer cell lines TK6, Jurkat, KG-1, and HCT116 (Stresemann et al.)
- Inhibits cell proliferation in T24 bladder carcinoma cells (Ben-Kasus et al.)
- In combination with the histone deacetylase inhibitor SAHA, reduces cell proliferation and increases apoptosis in pancreatic cancer cell lines (Neureiter et al.)
- Inhibits cell proliferation and induces apoptosis in human acute myeloid leukemia cells in vitro (Scott et al.)
- Increases the proportion of cells with cancer stem cell properties in subpopulations of human cancer cells and liver cancer cell lines (Marquardt et al.)
Tekniske data
Tekniske data
| CAS | 3690-10-6 |
| Synonym | NSC 309132 |
| Mængde | 10 mg |
| Molekylær formel | C9H12N2O5 |
| Renhed | ≥98% |
Produkttitel
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.
Ser du en mulighed for forbedring?