Få mere at vide
Zearalenone 4-Sulfate Ammonium Salt, TRC
Artikelnummer. 30451560
Skift visning
Mængde:
2.5 mg
25 mg
Pakningsstørrelse:
2.5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30451560 2.5 mg 2.5mg
30454300 25 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30451560 Leverandør Toronto Research Chemicals Leverandørnr. TRCZ2705302.5MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1439328-85-4
Molekylær formel C18H25NO8S
Molekylvægt (g/mol) 415.46
SMIL O=C(C1=C(O)C=C(OS(=O)([O-])=O)C=C1/C=C/CCC2)O[C@@H](C)CCCC2=O.[NH4+]
Tekniske data
Kemisk navn eller materiale Zearalenone 4-Sulfate Ammonium Salt
Mængde 2.5 mg
InChI formel InChI=1S/C18H22O8S.H3N/c1-12-6-5-9-14(19)8-4-2-3-7-13-10-15(26-27(22,23)24)11-16(20)17(13)18(21)25-12;/h3,7,10-12,20H,2,4-6,8-9H2,1H3,(H,22,23,24);1H3/b7-3+;/t12-;/m0./s1