missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
ZBTB8B Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-83789-0.1ML
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
ZBTB8B Polyclonal specifically detects ZBTB8B in Human samples. It is validated for Western Blot.
Tekniske data
| ZBTB8B | |
| Polyclonal | |
| PBS, 2% Sucrose | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 728116 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot | |
| Unconjugated | |
| DKFZp547H154, RP1-27O5.1, zinc finger and BTB domain containing 8B, zinc finger and BTB domain-containing protein 8B, ZNF916B | |
| The immunogen is a synthetic peptide directed towards the C-terminal region of Human ZBTB8B | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion