missing translation for 'onlineSavingsMsg'
Learn More

Zanamivir, MedChemExpress

Product Code. 16426116
Encompass_Preferred
Change view
Click to view available options
Mængde:
10 mg
50 mg
100 mg
Unit Size:
100mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde unitSize
16426116 100 mg 100mg
16425558 10 mg -
16411727 50 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16426116 Supplier Medchem Express Supplier No. HY13210

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Zanamivir is an influenza viral neuraminidase inhibitor with IC50 values of 0.95 nM and 2.7 nM for influenza A and B, respectively.

Zanamivir is an influenza viral neuraminidase inhibitor with IC50 values of 0.95 nM and 2.7 nM for influenza A and B, respectively.

Specifications

Kemisk navn eller materiale Zanamivir
CAS 139110-80-8
Farve White
Renhedsgrad noter Research
Molekylær formel C12H20N4O7
Mængde 100 mg
Opløselighedsinformation H2O : ≥ 33.33 mg/mL (100.30 mM)
SMIL NC(N[C@@H]1[C@H]([C@@]([H])(OC(C(O)=O)=C1)[C@@H]([C@@H](CO)O)O)NC(C)=O)=N
Molekylvægt (g/mol) 332.31
Formel vægt 332.31
Procent renhed 98.0%
Grad Research
Fysisk form Solid
Til brug med (applikation) COVID-19-anti-virus
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.