Beskrivelse
The ZA20D3 Overexpression Lysate is derived from human. It has been validated for the following applications: Western Blot.
Tekniske data
Tekniske data
| Cellelinje | The lysate was created in HEK293T cells, using plasmid ID RC22725 and based on accession number NM_196. The protein contains a C-terminal DDK tag. |
| Mængde | 0.1 mg |
| Opbevaringsbuffer | RIPA buffer |
| Renhed | Protein |
| Testspecificitet | Homo sapiens zinc finger, AN1-type domain 6 (ZFAND6), mRNA. |
| Indhold og opbevaring | -80°C. Avoid freeze-thaw cycles. |
| Produktlinje | Novus |
| Proteintilstand | Native |
| Til brug med (applikation) | Western Blot |
For Research Use Only