missing translation for 'onlineSavingsMsg'
Learn More

(Z)-Octa-1,5-dien-3-one (~0.1% Hydroquinone), TRC

Product Code. 30409026
Change view
Click to view available options
Mængde:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30409026 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30409026 Supplier Toronto Research Chemicals Supplier No. TRCO235830100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale (Z)-Octa-1,5-dien-3-one
CAS 65767-22-8
Molekylær formel C8H12O
Mængde 100 mg
Synonym (Z)-1,5-Octadien-3-one, cis-1,5-Octadien-3-one, (5Z)-1,5-Octadien-3-one
InChI formel InChI=1S/C8H12O/c1-3-5-6-7-8(9)4-2/h4-6H,2-3,7H2,1H3/b6-5-
SMIL C=CC(C/C=C\CC)=O
IUPAC navn (5Z)-octa-1,5-dien-3-one
Molekylvægt (g/mol) 124.18
Formel vægt 124.0888
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.