missing translation for 'onlineSavingsMsg'
Få mere at vide

STEMCELL Technologies (Z)-Guggulsterone

Artikelnummer. 17179971
Encompass_Preferred
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17179971 10 mg
17189971 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 17179971 Leverandør STEMCELL Technologies Leverandørnr. 1000252

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

Retinoic acid receptor (RAR) pathway inhibitor; Inhibits farnesoid X receptor (FXR)

  • (Z)-Guggulsterone is a plant steroid found in the resin of the guggul plant Commiphora mukul that acts as a selective antagonist of farnesoid X receptor (FXR; Cui et al.) It decreases chenodeoxycholic acid-induced FXR activation (IC50 = 10 μM) in the presence of 100 μM chenodeoxycholic acid (Cui et al.; Urizar et al.)
  • Induces differentiation of human pluripotent stem cell-derived neural stem cells into dopaminergic neurons (Gonzalez et al.)
  • Decreases hepatic low-density lipoprotein cholesterol and triglyceride levels in rodents fed with a high-cholesterol diet (Urizar et al.)
  • Regulates growth and metastasis of tumor cells in human multiple myeloma cells by the expression of STAT3-regulated antiapoptotic (Bcl-2, Bcl-xL, and Mcl-1), proliferative (cyclin D1), and angiogenic (VEGF) gene products (Ahn et al.)
  • Inhibits tube formation in human umbilical vein endothelial cells (HUVECs) and migration in HUVECs or a human prostate cancer cell line (Xiao et al.)

Tekniske data

CAS 39025-23-5
Mængde 10 mg
Molekylær formel C21H28O2
Renhed ≥98%
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.