Få mere at vide
Z-[4-(1,2-Diphenyl-1-butenyl)phenoxy]acetic Acid, TRC
Artikelnummer. 30133671
Skift visning
Mængde:
50 mg
250 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30133671 500 mg
30011913 250 mg
30008445 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30133671 Leverandør Toronto Research Chemicals Leverandørnr. TRCD489468500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 97818-93-4
Molekylær formel C24H22O3
Molekylvægt (g/mol) 358.43
Synonym Carboxy-tamoxifen
IUPAC navn 2-[4-[(Z)-1,2-diphenylbut-1-enyl]phenoxy]acetic acid
SMIL CC\C(=C(/c1ccccc1)\c2ccc(OCC(=O)O)cc2)\c3ccccc3
Tekniske data
Kemisk navn eller materiale Z-[4-(1,2-Diphenyl-1-butenyl)phenoxy]acetic Acid
Mængde 500 mg
InChI formel InChI=1S/C24H22O3/c1-2-22(18-9-5-3-6-10-18)24(19-11-7-4-8-12-19)20-13-15-21(16-14-20)27-17-23(25)26/h3-16H,2,17H2,1H3,(H,25,26)/b24-22-
Formel vægt 358.1569