missing translation for 'onlineSavingsMsg'
Få mere at vide

(Z)-2-Phenylbut-2-enyl 3,4,5-Trimethoxybenzoate, Mikromol™

Artikelnummer. 30458147
Change view
Click to view available options
Mængde:
25 mg
100 mg
4 x 25 mg
Product Code. Mængde
30458147 4 x 25 mg
30049556 25 mg
30050188 100 mg
3 options
This item is not returnable. View return policy

Product Code. 30458147

Brand: Mikromol MM0417.1500254AMP

Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

(Z)-2-Phenylbut-2-enyl 3,4,5-Trimethoxybenzoate, Mikromol
TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale (Z)-2-Phenylbut-2-enyl 3,4,5-Trimethoxybenzoate
Analyt- eller komponentnavne (Z)-2-Phenylbut-2-enyl 3,4,5-Trimethoxybenzoate
Forsendelsestilstand Room Temperature
Molekylær formel C20 H22 O5
Certificeringer/compliance ISO 9001
Synonym (Z)-2-Phenylbut-2-enyl 3,4,5-Trimethoxybenzoate
InChI formel InChI=1S/C20H22O5/c1-5-14(15-9-7-6-8-10-15)13-25-20(21)16-11-17(22-2)19(24-4)18(12-16)23-3/h5-12H,13H2,1-4H3/b14-5+
Løsningstype Neat
SMIL COc1cc(cc(OC)c1OC)C(=O)OC\C(=C/C)\c2ccccc2
IUPAC navn [(Z)-2-phenylbut-2-enyl] 3,4,5-trimethoxybenzoate
Molekylvægt (g/mol) 342.39
Mængde 4 x 25 mg
Formel vægt 342.1467 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.