Få mere at vide
(Z)-2-(Methoxyimino)-3-oxobutanoic Acid-d3 Methyl Ester, TRC
Artikelnummer. 30468155
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30468155 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30468155 Leverandør Toronto Research Chemicals Leverandørnr. TRCM26268710MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale (Z)-2-(Methoxyimino)-3-oxobutanoic Acid-d3 Methyl Ester
CAS 1331636-65-7
Molekylær formel C6H6D3NO4
Mængde 10 mg
Synonym Methyl (Z)-2-Methoxyimino-3-oxobutanoate-d3
InChI formel InChI=1S/C6H9NO4/c1-4(8)5(7-11-3)6(9)10-2/h1-3H3/b7-5-/i3D3
SMIL [2H]C([2H])([2H])O\N=C(\C(=O)C)/C(=O)OC
IUPAC navn methyl (2Z)-3-oxo-2-(trideuteriomethoxyimino)butanoate
Molekylvægt (g/mol) 162.16
Formel vægt 162.072