missing translation for 'onlineSavingsMsg'
Få mere at vide

(Z)-2-Methoxyimino-2-furanacetic Acid Ammonium Salt, TRC

Artikelnummer. 30467052
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30467052 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30467052 Leverandør Toronto Research Chemicals Leverandørnr. TRCM26237525G

Please to purchase this item. Need a web account? Register with us today!

Denne vare kræver en prisopdatering, inden den kan bestilles.

Anmod om en pris på denne vare her.

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Tekniske data

Kemisk navn eller materiale (Z)-2-Methoxyimino-2-furanacetic acid ammonium salt
CAS 97148-39-5
Molekylær formel C7 H6 N O4 . H4 N
Mængde 25 g
Synonym 2-Furanacetic acid, α-(methoxyimino)-, ammonium salt (1:1), (αZ)-, 2-Furanacetic acid, α-(methoxyimino)-, ammonium salt, (Z)-, 2-Furanacetic acid, α-(methoxyimino)-, ammonium salt, (αZ)- (9CI), Ammonium (Z)-(Furan-2-yl)(methoxyimino)acetate, Cefuroxime sodium related, 640-ammonium salt, 750-acid (anti isomer), 736-acid (furyl glyoxylic acid), 2-furoic acid
InChI formel InChI=1S/C7H7NO4.H3N/c1-11-8-6(7(9)10)5-3-2-4-12-5;/h2-4H,1H3,(H,9,10);1H3/b8-6-;
SMIL [NH4+].CO\N=C(/C(=O)[O-])\c1occc1
IUPAC navn azanium;(2Z)-2-(furan-2-yl)-2-methoxyiminoacetate
Molekylvægt (g/mol) 186.17
Formel vægt 186.0641
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.