Få mere at vide
(Z)-2-Chloropropane-1,3-diyl Dioleate, TRC
Artikelnummer. 17991785
Skift visning
Mængde:
25 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17991785 250 mg
17972435 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 17991785 Leverandør Toronto Research Chemicals Leverandørnr. TRCC380305250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1639207-42-3
Molekylær formel C39 H71 Cl O4
Molekylvægt (g/mol) 639.43
IUPAC navn [2-chloro-3-[(Z)-octadec-9-enoyl]oxy-propyl] (Z)-octadec-9-enoate
SMIL CCCCCCCC\C=C/CCCCCCCC(=O)OCC(Cl)COC(=O)CCCCCCC\C=C/CCCCCCCC
Tekniske data
Kemisk navn eller materiale (Z)-2-Chloropropane-1,3-diyl Dioleate
Mængde 250 mg
InChI formel InChI=1S/C39H71ClO4/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-31-33-38(41)43-35-37(40)36-44-39(42)34-32-30-28-26-24-22-20-18-16-14-12-10-8-6-4-2/h17-20,37H,3-16,21-36H2,1-2H3/b19-17-,20-18-
Formel vægt 638.5041