Få mere at vide
(Z)-13-Octadecenal, TRC
Artikelnummer. 30008217
Skift visning
Mængde:
2.5 mg
10 mg
25 mg
Pakningsstørrelse:
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30008217 25 mg -
17910445 2.5 mg 2.5mg
17932605 10 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30008217 Leverandør Toronto Research Chemicals Leverandørnr. TRCO23550025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 58594-45-9
Molekylær formel C18H34O
Molekylvægt (g/mol) 266.46
Synonym (13Z)-13-Octadecenal, cis-13-Octadecenal;
IUPAC navn (Z)-octadec-13-enal
SMIL CCCC\C=C/CCCCCCCCCCCC=O
Tekniske data
Kemisk navn eller materiale (Z)-13-Octadecenal
Mængde 25 mg
InChI formel InChI=1S/C18H34O/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19/h5-6,18H,2-4,7-17H2,1H3/b6-5-
Formel vægt 266.261