missing translation for 'onlineSavingsMsg'
Learn More

(Z)-11-Octadecen-1-ol, TRC

Product Code. 30417532
Change view
Click to view available options
Mængde:
50 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30417532 500 mg
30416814 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30417532 Supplier Toronto Research Chemicals Supplier No. TRCO235570500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale (Z)-11-Octadecen-1-ol
CAS 62972-93-4
Molekylær formel C18 H36 O
Mængde 500 mg
Synonym (Z)-11-Octadecen-1-ol
InChI formel InChI=1S/C18H36O/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19/h7-8,19H,2-6,9-18H2,1H3/b8-7-
SMIL CCCCCC\C=C/CCCCCCCCCCO
IUPAC navn (Z)-octadec-11-en-1-ol
Molekylvægt (g/mol) 268.478
Formel vægt 268.277
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.