Få mere at vide
(Z)-11-Octadecen-1-ol, TRC
Artikelnummer. 30416814
Skift visning
Mængde:
50 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30416814 50 mg
30417532 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30416814 Leverandør Toronto Research Chemicals Leverandørnr. TRCO23557050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale (Z)-11-Octadecen-1-ol
CAS 62972-93-4
Molekylær formel C18 H36 O
Mængde 50 mg
Synonym (Z)-11-Octadecen-1-ol
InChI formel InChI=1S/C18H36O/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19/h7-8,19H,2-6,9-18H2,1H3/b8-7-
SMIL CCCCCC\C=C/CCCCCCCCCCO
IUPAC navn (Z)-octadec-11-en-1-ol
Molekylvægt (g/mol) 268.478
Formel vægt 268.277