Få mere at vide
(Z)-11-Hexadecen-1-ol Acetate, TRC
Artikelnummer. 30180187
Skift visning
Mængde:
250 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30180187 500 mg
30180154 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30180187 Leverandør Toronto Research Chemicals Leverandørnr. TRCH293685500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 34010-21-4
Molekylær formel C18 H34 O2
Molekylvægt (g/mol) 282.46
Synonym 11-Hexadecen-1-ol, acetate, (11Z)- (9CI), 11-Hexadecen-1-ol, acetate, (Z)- (8CI), (Z)-11-Hexadecen-1-ol acetate, (Z)-11-Hexadecen-1-yl acetate, (Z)-11-Hexadecen-1-yl acetate, (Z)-11-Hexadecenyl acetate, 1-Acetoxy-(Z)-11-hexadecene, Z11-16:Ac, cis-11-Hexadecen-1-ol acetate, cis-11-Hexadecen-1-yl acetate, cis-11-Hexadecenyl acetate
IUPAC navn [(Z)-hexadec-11-enyl] acetate
SMIL CCCC\C=C/CCCCCCCCCCOC(=O)C
Tekniske data
Kemisk navn eller materiale (Z)-11-Hexadecen-1-yl acetate
Mængde 500 mg
InChI formel InChI=1S/C18H34O2/c1-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-20-18(2)19/h6-7H,3-5,8-17H2,1-2H3/b7-6-
Formel vægt 282.2559