missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
YY1 Rabbit anti-Human, Mouse, Rat, Clone: S02-8A3, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Bio-Techne NBP3-14987-100UL
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
YY1 Monoclonal antibody specifically detects YY1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence
Tekniske data
| YY1 | |
| Monoclonal | |
| Unconjugated | |
| DELTA, Delta transcription factor, INO80SYY-1, NF-E1INO80 complex subunit S, transcriptional repressor protein YY1, UCRBP, Yin and yang 1, Yin and Yang 1 protein, YIN-YANG-1, YY1 transcription factor | |
| A synthetic peptide of human YY1 (Uniprot#: P25490) | |
| 100 μg | |
| Chromatin Research, Core ESC Like Genes, Stem Cell Markers | |
| 7528 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry, Immunofluorescence | |
| S02-8A3 | |
| 50nM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA with 0.01% Sodium Azide | |
| Rabbit | |
| Affinity-purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion