Få mere at vide
Thermo Scientific™ Yttria Stabilized Zirconium Oxide Conical Crucibles, High Form
Kapacitet (metrisk):
50 ml
100 ml
200 ml
Diameter (metrisk) Udvendig:
51 mm
62 mm
77 mm
Højde (metrisk):
58 mm
72 mm
90 mm
Pakningsstørrelse:
Hver
Beskrivelse
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Tekniske data
Tekniske data
| Materiale | Zirconium Oxide |
| Mængde | 1 Piece |
| Kapacitet (metrisk) | 200 ml |
| Diameter (metrisk) Udvendig | 77 mm |
| Højde (metrisk) | 90 mm |
| Produkttype | Yttria Stabilized Zirconium Oxide Conical Crucibles, High Form |
| Diameter (metrisk) base | 50mm |