Få mere at vide
YH239-EE, MedChemExpress
Artikelnummer. 16435068
Encompass_Preferred
Skift visning
Mængde:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
16435068 10 mg
16401307 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 16435068 Leverandør Medchem Express Leverandørnr. HY12287
MedChemExpress YH239-EE, ethyl ester of the free carboxylic acid compound YH239, is a potent p53-MDM2 antagonizing and apoptosis-inducing agent.
YH239-EE, ethyl ester of the free carboxylic acid compound YH239, is a potent p53-MDM2 antagonizing and apoptosis-inducing agent.
Tekniske data
Kemisk navn eller materiale YH239-EE
Fysisk form Solid
CAS 1364488-67-4
Farve Off-White
Mængde 10 mg
Renhedsgrad noter Research
Molekylær formel C25H27Cl2N3O4
Til brug med (applikation) Cancer-programmed cell death
Opløselighedsinformation DMSO : ≥ 55 mg/mL (109.04 mM)
SMIL O=C(C(N1)=C(C(N(CC2=CC=C(Cl)C=C2)C=O)C(NC(C)(C)C)=O)C3=C1C=C(Cl)C=C3)OCC
Molekylvægt (g/mol) 504.41
Formel vægt 504.41
Procent renhed 99.83%