Få mere at vide
Tocris Bioscience™ YC 1
GREENER_CHOICE
Artikelnummer. 18065692
Skift visning
Mængde:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
18065692 50 mg
18064852 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 18065692 Leverandør Tocris Bioscience™ Leverandørnr. 4307/50
Soluble guanylyl cyclase (sGC) activator; induces G1 cell cycle arrest
Nitric oxide-independent activator of soluble guanylyl cyclase (sGC). Significantly elevates cGMP levels and inhibits collagen-stimulated aggregation of washed rabbit platelets (IC50 = 14.6 μM); induces relaxation in denuded phenylephrine-contracted rabbit aortic rings (EC50 = 1.9 μM). Also displays antiproliferative activity in vitro and in vivo by inducing G1 cell cycle arrest in two human hepatocellular carcinoma (HCC) cell lines, and in HCC xenografts in athymic SCID mice. Exhibits low cytotoxicity in non-malignant cells.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 170632-47-0
Molekylær formel C19H16N2O2
Tekniske data
Kemisk navn eller materiale YC 1
Mængde 50 mg
Formel vægt 304.34
Renhed >99%
Molekylvægt (g/mol) 304.35